Learn More
Medchemexpress LLC Isocurcumenol | 24063-71-6 | 5 MG
SDP
Catalog No. 5000421054
Encompass_Preferred
Isocurcumenol is an estrogen receptor alpha (ERα) inhibitor isolated from Curcuma zedoaria Rhizomes. It possesses anti-tumor activity, with IC50 values of 99.1 μg/mL and 178.2 μg/mL in DLA and KB cells, respectively. In vitro, Isocurcumenol dose-dependently inhibits cancer cell proliferation and does not exhibit significant toxicity on normal chicken embryo fibroblast cells and lymphocytes.
  • ERα inhibitor
  • Isolated from Curcuma zedoaria Rhizomes
  • Possesses anti-tumor activity
  • Inhibits cancer cell proliferation in a dose-dependent manner
  • Does not show significant toxicity on normal chicken embryo fibroblast cells and lymphocytes
Catalog No. 50-004-21054 Supplier Medchemexpress LLC Supplier No. HYN41215MG
Isocurcumenol is an estrogen receptor alpha (ERα) inhibitor isolated from Curcuma zedoaria Rhizomes. It possesses anti-tumor activity, with IC50 values of 99.1 μg/mL and 178.2 μg/mL in DLA and KB cells, respectively. In vitro, Isocurcumenol dose-dependently inhibits cancer cell proliferation and does not exhibit significant toxicity on normal chicken embryo fibroblast cells and lymphocytes.
  • ERα inhibitor
  • Isolated from Curcuma zedoaria Rhizomes
  • Possesses anti-tumor activity
  • Inhibits cancer cell proliferation in a dose-dependent manner
  • Does not show significant toxicity on normal chicken embryo fibroblast cells and lymphocytes