Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isocurcumenol | 24063-71-6 | MFCD03792802 | 99.5% | 234.33 g/mol | C15H22O2 | 10 MG
SDP

Catalog No. 5000289794
Encompass_Preferred

Isocurcumenol is a sesquiterpene terpenoid isolated from Curcuma zedoaria rhizomes that acts as an estrogen receptor alpha (ERα) inhibitor and shows reported anti-tumor activity. Supplied as a white to off-white solid for research use only, it has molecular formula C15H22O2 and molecular weight 234.33 g/mol.

  • High purity (99.47%).
  • White to off-white solid physical form.
  • Soluble in DMSO (100 mg/mL); in vivo vehicle example: 10% DMSO, 90% corn oil.
  • Recommended storage: -20°C for solid; protect from light; solutions: -80°C up to 6 months.

Catalog No. 50-002-89794 Supplier Medchemexpress LLC Supplier No. HYN412110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isocurcumenol is a sesquiterpene terpenoid isolated from Curcuma zedoaria rhizomes that acts as an estrogen receptor alpha (ERα) inhibitor and shows reported anti-tumor activity. Supplied as a white to off-white solid for research use only, it has molecular formula C15H22O2 and molecular weight 234.33 g/mol.

  • High purity (99.47%).
  • White to off-white solid physical form.
  • Soluble in DMSO (100 mg/mL); in vivo vehicle example: 10% DMSO, 90% corn oil.
  • Recommended storage: -20°C for solid; protect from light; solutions: -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.