Learn More
Medchemexpress LLC Isocytosine | 108-53-2 | 111.10 | 10 G
SDP
Catalog No. 5000449564
Encompass_Preferred
Isocytosine is a non-natural nucleobase, an isomer of cytosine. It is used in combination with Isoguanine for research into unnatural nucleic acid analogues of the normal base pairs found in DNA, and also serves as a nucleobase of hachimoji RNA.
  • Used for research into unnatural nucleic acid analogues
  • Serves as a nucleobase of hachimoji RNA
  • Has a purity of 99.6%
  • Molecular weight of 111.10
  • Appears as an off-white to light yellow solid
Catalog No. 50-004-49564 Supplier Medchemexpress LLC Supplier No. HYW00227210G
Isocytosine is a non-natural nucleobase, an isomer of cytosine. It is used in combination with Isoguanine for research into unnatural nucleic acid analogues of the normal base pairs found in DNA, and also serves as a nucleobase of hachimoji RNA.
  • Used for research into unnatural nucleic acid analogues
  • Serves as a nucleobase of hachimoji RNA
  • Has a purity of 99.6%
  • Molecular weight of 111.10
  • Appears as an off-white to light yellow solid