Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isocytosine | 108-53-2 | 111.10 | 25 G
SDP

Catalog No. 5000448265
Encompass_Preferred

Isocytosine is a non-natural nucleobase and an isomer of cytosine. It is used in combination with Isoguanine in studies of unnatural nucleic acid analogues of the normal base pairs in DNA and as a nucleobase of hachimoji RNA.

  • Non-natural nucleobase
  • Isomer of cytosine
  • Used in combination with Isoguanine in studies
  • Applied in unnatural nucleic acid analogues of normal base pairs in DNA
  • Functions as a nucleobase of hachimoji RNA
  • Molecular weight: 111.10
  • Formula: C₄H₅N₃O
  • CAS number: 108-53-2
  • Purity: 99.7%
  • Appearance: Solid
  • Color: Off-white to light yellow

Catalog No. 50-004-48265 Supplier Medchemexpress LLC Supplier No. 25GHYW002272
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isocytosine is a non-natural nucleobase and an isomer of cytosine. It is used in combination with Isoguanine in studies of unnatural nucleic acid analogues of the normal base pairs in DNA and as a nucleobase of hachimoji RNA.

  • Non-natural nucleobase
  • Isomer of cytosine
  • Used in combination with Isoguanine in studies
  • Applied in unnatural nucleic acid analogues of normal base pairs in DNA
  • Functions as a nucleobase of hachimoji RNA
  • Molecular weight: 111.10
  • Formula: C₄H₅N₃O
  • CAS number: 108-53-2
  • Purity: 99.7%
  • Appearance: Solid
  • Color: Off-white to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.