Learn More
Medchemexpress LLC Isocytosine | 108-53-2 | 111.10 | 50 G
SDP
Catalog No. 5000445911
Encompass_Preferred
Isocytosine is a non-natural nucleobase that is an isomer of cytosine. It is utilized in research involving unnatural nucleic acid analogues, specifically in combination with Isoguanine, to study alternatives to the normal base pairs found in DNA. Additionally, it serves as a nucleobase component of hachimoji RNA.
  • Non-natural nucleobase
  • Isomer of cytosine
  • Used with isoguanine in unnatural nucleic acid analogue studies
  • Component of hachimoji RNA
  • Appearance: solid
  • Color: off-white to light yellow
Catalog No. 50-004-45911 Supplier Medchemexpress LLC Supplier No. HYW00227250G
Isocytosine is a non-natural nucleobase that is an isomer of cytosine. It is utilized in research involving unnatural nucleic acid analogues, specifically in combination with Isoguanine, to study alternatives to the normal base pairs found in DNA. Additionally, it serves as a nucleobase component of hachimoji RNA.
  • Non-natural nucleobase
  • Isomer of cytosine
  • Used with isoguanine in unnatural nucleic acid analogue studies
  • Component of hachimoji RNA
  • Appearance: solid
  • Color: off-white to light yellow