Learn More
Apexbio Technology LLC Isocytosine 108-53-2 200mg
SDP
Catalog No. 5000090076
Encompass_Preferred
Isocytosine (CAS 108-53-2) is a non-canonical nucleobase and a structural isomer of cytosine It serves as an analog in nucleic acid research notably as a component of expanded genetic systems such as hachimoji RNA and can pair with isoguanine to model non-standard base pairing Mechanistically isocytosine has been shown to modulate specific signaling pathways and enzyme activities in experimental systems It is widely utilized in molecular biology and genetics to investigate alternative base-pairing nucleic acid stability and the physicochemical properties of synthetic genetic polymers
Catalog No. 50-000-90076 Supplier Apexbio Technology LLC Supplier No. C7013200
Isocytosine (CAS 108-53-2) is a non-canonical nucleobase and a structural isomer of cytosine It serves as an analog in nucleic acid research notably as a component of expanded genetic systems such as hachimoji RNA and can pair with isoguanine to model non-standard base pairing Mechanistically isocytosine has been shown to modulate specific signaling pathways and enzyme activities in experimental systems It is widely utilized in molecular biology and genetics to investigate alternative base-pairing nucleic acid stability and the physicochemical properties of synthetic genetic polymers