Learn More
Medchemexpress LLC Isodecyl diphenyl phosphate | 29761-21-5 | MFCD00072254 | 99.0% | 390.45 | C22H31O4P | 10mg
SDP
Catalog No. 5000270125
Encompass_Preferred
Isodecyl diphenyl phosphate (IDPP) is a flame retardant IDPP has an ability to induce DNA damage and exertes S phase cell ratio-reducing effects in A549 cells[1]
Catalog No. 50-002-70125 Supplier Medchemexpress LLC Supplier No. HYW71842310MG
Isodecyl diphenyl phosphate (IDPP) is a flame retardant IDPP has an ability to induce DNA damage and exertes S phase cell ratio-reducing effects in A549 cells[1]