Learn More
Medchemexpress LLC Isodecyl diphenyl phosphate | 29761-21-5 | MFCD00072254 | 99.0% | 390.45 g/mol | C22H31O4P | 50mg
SDP
Catalog No. 5000269768
Encompass_Preferred
Isodecyl diphenyl phosphate (IDPP) is a flame retardant IDPP has an ability to induce DNA damage and exertes S phase cell ratio-reducing effects in A549 cells[1]
Catalog No. 50-002-69768 Supplier Medchemexpress LLC Supplier No. HYW71842350MG
Isodecyl diphenyl phosphate (IDPP) is a flame retardant IDPP has an ability to induce DNA damage and exertes S phase cell ratio-reducing effects in A549 cells[1]