Learn More
Cayman Chemical Isodeoxycholic Acid, Purity: ≥95%, CAS: 566-17-6, 10 mg
SDP
Catalog No. 502989254
Encompass_Preferred
A bile acid and an FXR agonist; induces FXR-mediated gene expression in a reporter assay using COS-7 cells at 30 µM; activates KCa1.1/BK in isolated rabbit superior mesenteric arteries
Catalog No. 50-298-9254 Supplier Cayman Chemical Supplier No. 2989010 MG
A bile acid and an FXR agonist; induces FXR-mediated gene expression in a reporter assay using COS-7 cells at 30 µM; activates KCa1.1/BK in isolated rabbit superior mesenteric arteries