Learn More
Medchemexpress LLC Isodeoxycholic acid (7α,12α-dihydroxycholanoic acid) | 566-17-6 | MFCD00036859 | 99.6% | 392.57 g/mol | C24H40O4 | 5 MG
SDP
Catalog No. 5000281333
Encompass_Preferred
Isodeoxycholic acid (7α,12α-dihydroxycholanoic acid) is a bile acid research reagent and analytical standard. It is the 3β-epimer of ursodeoxycholic acid and is used to study choleresis, liver biochemistry, and bile acid metabolism. The compound is supplied as a high-purity solid and is readily soluble in DMSO for solution preparation.
  • High purity (99.62%) solid suitable for analytical and biochemical studies.
  • Useful as a research standard for bile acid profiling and metabolism studies.
  • Supplied in small quantities appropriate for research-scale experiments.
  • Stable under recommended storage conditions for extended shelf life.
  • Readily soluble in DMSO, enabling easy reconstitution for assays.
Catalog No. 50-002-81333 Supplier Medchemexpress LLC Supplier No. HYW3992975MG
Add to Cart
Add to Cart
Isodeoxycholic acid (7α,12α-dihydroxycholanoic acid) is a bile acid research reagent and analytical standard. It is the 3β-epimer of ursodeoxycholic acid and is used to study choleresis, liver biochemistry, and bile acid metabolism. The compound is supplied as a high-purity solid and is readily soluble in DMSO for solution preparation.
  • High purity (99.62%) solid suitable for analytical and biochemical studies.
  • Useful as a research standard for bile acid profiling and metabolism studies.
  • Supplied in small quantities appropriate for research-scale experiments.
  • Stable under recommended storage conditions for extended shelf life.
  • Readily soluble in DMSO, enabling easy reconstitution for assays.