Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Isodesmosine

Catalog No. MP219137905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219137905 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219137905 Supplier MP Biomedicals™ Supplier No. 0219137905
Add to Cart
Add to Cart

Isodesmosine from Bovine Neck Ligament≥99%, Yellow Solid

  • Isodesmosine is prepared according to the method of Starcher, et al. from hdrolyzed bovine neck ligament by solvent extraction and chromatography on Dowex 50.
  • From Bovine Neck Ligament
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 526
Quantity 5 mg
Source Bovine Neck Ligament
Storage Requirements Store at +4°C, desiccate
Product Type Biochemicals
Form Solid
Purity or Quality Grade > 99% (amino acid analysis)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.