Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isodesmosine, ≥99%, MP Biomedicals™

Catalog No. ICN19137910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19137910 10 mg
ICN19137901 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19137910 Supplier MP Biomedicals, Inc Supplier No. 0219137910
Add to Cart
Add to Cart
Request a Quote
Molecular biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 991-01-5
Molecular Formula C24H40N5O8
Molecular Weight (g/mol) 526.61
InChI Key RGXCTRIQQODGIZ-UHFFFAOYNA-O
Synonym isodesmosine, 2-amino-6-3,5-bis 3-amino-4-hydroxy-4-oxobutyl-2-4-amino-5-hydroxy-5-oxopentyl pyridin-1-ium-1-yl hexanoic acid, iso-desmosin, 6-2-4-amino-4-carboxybutyl-3,5-bis 3-amino-3-carboxypropyl pyridinium-1-yl norleucine, 2-4-amino-4-carboxybutyl-1-5-amino-5-carboxypentyl-3,5-bis 3-amino-3-carboxypropyl-pyridinium, 2-4-amino-4-carboxybutyl-1-5-amino-5-carboxypentyl-3,5-bis 3-amino-3-carboxypropyl pyridinium, 3,5-bis 3-amino-3-carboxypropyl-2-4-amino-4-carboxybutyl-1-5-amino-5-carboxypentyl pyridin-1-ium, pyridinium, 2-4-amino-4-carboxybutyl-1-5-amino-5-carboxypentyl-3,5-bis 3-amino-3-carboxypropyl
PubChem CID 13811
ChEBI CHEBI:64366

Specifications

Color Yellow
Assay Percent Range ≥99%
Quantity 10 mg
Formula Weight 526
Physical Form Solid
Chemical Name or Material Isodesmosine
storageNote1 Store Desiccate.
Recommended Storage Store at +4°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.