Learn More
Medchemexpress LLC Isoeugenol | 97-54-1 | 164.20 | 10 G
SDP
Catalog No. 5000415818
Encompass_Preferred
Isoeugenol is an essential oil constituent found in nutmeg, clove, and cinnamon. It inhibits the growth of *Escherichia coli* and *Listeria innocua*.
  • Inhibits various Gram-positive and Gram-negative bacteria
  • Used for research purposes
  • Available in liquid form
Catalog No. 50-004-15818 Supplier Medchemexpress LLC Supplier No. HYN195210G
Isoeugenol is an essential oil constituent found in nutmeg, clove, and cinnamon. It inhibits the growth of *Escherichia coli* and *Listeria innocua*.
  • Inhibits various Gram-positive and Gram-negative bacteria
  • Used for research purposes
  • Available in liquid form