Learn More
TARGETMOL CHEMICALS INC Isoeugenol acetate 1G
SDP
Catalog No. 502878871
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 1. Isoeugenol acetate (Acetylisoeugenol) shows good antifungal activities against R. solani and F. oxysporum. 2. Isoeugenol acetate is a mild sensitizer in vivo. Purity 99.76%
Catalog No. 50-287-8871 Supplier TARGETMOL CHEMICALS INC Supplier No. T2S04911G
Also available in 1 mL, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 1. Isoeugenol acetate (Acetylisoeugenol) shows good antifungal activities against R. solani and F. oxysporum. 2. Isoeugenol acetate is a mild sensitizer in vivo. Purity 99.76%