Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isoeugenol acetate | 93-29-8 | 100 MG
SDP

Catalog No. 5000417798
Encompass_Preferred

Isoeugenol acetate, also known as Acetyl isoeugenol, is a derivative of Isoeugenol that acts as an inhibitor for ferroptosis and targets AChE. In vitro studies demonstrate its ability to inhibit cell death and alleviate ROS accumulation and lipid peroxidation in stimulated HT-1080 cells. This product is for research use only and has not been fully validated for medical applications.

  • Inhibitor for ferroptosis
  • Targets AChE
  • Inhibits cell death
  • Alleviates ROS accumulation and lipid peroxidation
  • Suitable for laboratory chemical applications
  • Solid appearance
  • Molecular weight of 206.24
  • Molecular formula C12H14O3

Catalog No. 50-004-17798 Supplier Medchemexpress LLC Supplier No. HYN6805100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoeugenol acetate, also known as Acetyl isoeugenol, is a derivative of Isoeugenol that acts as an inhibitor for ferroptosis and targets AChE. In vitro studies demonstrate its ability to inhibit cell death and alleviate ROS accumulation and lipid peroxidation in stimulated HT-1080 cells. This product is for research use only and has not been fully validated for medical applications.

  • Inhibitor for ferroptosis
  • Targets AChE
  • Inhibits cell death
  • Alleviates ROS accumulation and lipid peroxidation
  • Suitable for laboratory chemical applications
  • Solid appearance
  • Molecular weight of 206.24
  • Molecular formula C12H14O3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.