Learn More
Medchemexpress LLC Isoeugenol acetate | 93-29-8 | 500 MG
SDP
Catalog No. 5000417762
Encompass_Preferred
Isoeugenol acetate (Acetyl isoeugenol) is a derivative of Isoeugenol and functions as an inhibitor for ferroptosis. In vitro, it inhibits cell death and reduces reactive oxygen species (ROS) accumulation and lipid peroxidation in RSL3 or IKE-stimulated HT-1080 cells. It also improved cell viability. The product has an appearance of a white to off-white solid and is sourced from plants like Annonaceae and Cananga odorata.
  • Inhibits cell death
  • Reduces reactive oxygen species (ROS) accumulation
  • Reduces lipid peroxidation in RSL3 or IKE-stimulated HT-1080 cells
  • Improves cell viability
  • Appears as a white to off-white solid
  • Derived from plants
Catalog No. 50-004-17762 Supplier Medchemexpress LLC Supplier No. HYN6805500MG
Isoeugenol acetate (Acetyl isoeugenol) is a derivative of Isoeugenol and functions as an inhibitor for ferroptosis. In vitro, it inhibits cell death and reduces reactive oxygen species (ROS) accumulation and lipid peroxidation in RSL3 or IKE-stimulated HT-1080 cells. It also improved cell viability. The product has an appearance of a white to off-white solid and is sourced from plants like Annonaceae and Cananga odorata.
  • Inhibits cell death
  • Reduces reactive oxygen species (ROS) accumulation
  • Reduces lipid peroxidation in RSL3 or IKE-stimulated HT-1080 cells
  • Improves cell viability
  • Appears as a white to off-white solid
  • Derived from plants