Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoeugenol, MilliporeSigma™ Supelco™

Catalog No. 111015993 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5993 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5993 Supplier MilliporeSigma Supelco Supplier No. 340385MLR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoeugenol belongs to the class of phenylpropenes, naturally occurring in the essential oils of plants. It can also be obtained from eugenol by isomerization. It has been used as a substrate in the preparation of vanillin via enzymatic transformation by lipoxygenase enzyme.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoeugenol
CAS 97-54-1
Vapor Pressure <0.01 mmHg (20°C)
Vapor Density >1 (vs. air)
Density 1.084 g/cm3
Flash Point 112°C - Closed Cup
Physical Form Neat
Linear Formula CH3 OC6H3(CH=CHCH3)OH
Technique HPLC
MDL Number MFCD00009285
Boiling Point 132°C (10 mmHg)
Synonym 2-Methoxy-4-propenylphenol
Type Analytical Standard
Industry Type Food and Beverage
Molecular Weight (g/mol) 164.20
Quantity 5 mL
Formula Weight 164.20
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.