Learn More
AdipoGen Isoeugenyl acetate
SDP
Catalog No. 502560630
Encompass_Preferred
Chemical. CAS 93-29-8. Formula C12H14O3. MW 206.2. Isoeugenol is used as a fragrance material in perfumes and perfumed products. It is a moderate to strong skin allergen and a frequent cause of contact allergy. Isoeugenyl acetate has been used as a substitute for Isoeugenol, but is probably easily hydrolysed in the skin to produce isoeugenol. Isoeugenyl acetate is a useful building block and has antibacterial and antifungal properties. Compound can be used as analytical reference material.
Catalog No. 50-256-0630 Supplier AdipoGen Supplier No. CDXI0198M250
Chemical. CAS 93-29-8. Formula C12H14O3. MW 206.2. Isoeugenol is used as a fragrance material in perfumes and perfumed products. It is a moderate to strong skin allergen and a frequent cause of contact allergy. Isoeugenyl acetate has been used as a substitute for Isoeugenol, but is probably easily hydrolysed in the skin to produce isoeugenol. Isoeugenyl acetate is a useful building block and has antibacterial and antifungal properties. Compound can be used as analytical reference material.