Learn More
Selleck Chemical LLC Isoferulic Acid S3764-5mg
SDP
Catalog No. 503110753
Encompass_Preferred
Isoferulic acid (Hesperetic acid Hesperetate Isoferulate) a major active ingredient of Cimicifuga heracleifolia shows anti-inflammatory antiviral antioxidant and antidiabetic properties Isoferulic acid (3-Hydroxy-4-methoxycinnamic acid) is a cinnamic acid derivative that has antidiabetic activity Isoferulic acid binds to and activates 1-adrenergic receptors (IC50 1 4 M) to enhance secretion of -endorphin (EC50 52 2 nM) and increase glucose use
Catalog No. 50-311-0753 Supplier Selleck Chemical LLC Supplier No. S37645MG
Add to Cart
Add to Cart
Isoferulic acid (Hesperetic acid Hesperetate Isoferulate) a major active ingredient of Cimicifuga heracleifolia shows anti-inflammatory antiviral antioxidant and antidiabetic properties Isoferulic acid (3-Hydroxy-4-methoxycinnamic acid) is a cinnamic acid derivative that has antidiabetic activity Isoferulic acid binds to and activates 1-adrenergic receptors (IC50 1 4 M) to enhance secretion of -endorphin (EC50 52 2 nM) and increase glucose use