Learn More
Medchemexpress LLC Isoflavone | 1374524-68-1 | 99.8% | 348.39 | 50 MG
SDP
Catalog No. 5000436586
Encompass_Preferred
ME-344 is an Isoflavone that increases mitochondrial ROS generation, inhibits tubulin polymerization, and inhibits HO-1 and impacts its mitochondrial translocation. It induces Apoptosis through Caspase 3 activation and synergizes with vinblastine in leukemia cells. This compound displays anti-tumor activity against leukemia and lung tumors and can be used in the research of lung cancer, acute myeloid leukemia, and HER2-negative breast cancer.
  • Increases mitochondrial ROS generation.
  • Inhibits tubulin polymerization.
  • Inhibits HO-1 and impacts its mitochondrial translocation.
  • Induces Apoptosis through Caspase 3 activation.
  • Synergizes with vinblastine in leukemia cells.
  • Displays anti-tumor activity against leukemia and lung tumor.
  • Used in research for lung cancer, acute myeloid leukemia, and HER2-negative breast cancer.
Catalog No. 50-004-36586 Supplier Medchemexpress LLC Supplier No. HY11274950MG
ME-344 is an Isoflavone that increases mitochondrial ROS generation, inhibits tubulin polymerization, and inhibits HO-1 and impacts its mitochondrial translocation. It induces Apoptosis through Caspase 3 activation and synergizes with vinblastine in leukemia cells. This compound displays anti-tumor activity against leukemia and lung tumors and can be used in the research of lung cancer, acute myeloid leukemia, and HER2-negative breast cancer.
  • Increases mitochondrial ROS generation.
  • Inhibits tubulin polymerization.
  • Inhibits HO-1 and impacts its mitochondrial translocation.
  • Induces Apoptosis through Caspase 3 activation.
  • Synergizes with vinblastine in leukemia cells.
  • Displays anti-tumor activity against leukemia and lung tumor.
  • Used in research for lung cancer, acute myeloid leukemia, and HER2-negative breast cancer.