Learn More
Medchemexpress LLC Isoflavone | 574-12-9 | 222.24 | 250 MG
SDP
Catalog No. 5000449888
Encompass_Preferred
Isoflavone is an orally available bioactive component of soy phytoestrogen with lipid-lowering and antioxidant activities. It prevents a variety of chronic diseases by regulating fatty acid oxidation in the liver and gene expression in adipose tissue. It is also valuable in cancer and cardiovascular disease research.
  • Orally available bioactive component
  • Regulates fatty acid oxidation in the liver
  • Regulates gene expression in adipose tissue
  • Useful in cancer and cardiovascular disease research
Catalog No. 50-004-49888 Supplier Medchemexpress LLC Supplier No. 250MGHYW006405
Isoflavone is an orally available bioactive component of soy phytoestrogen with lipid-lowering and antioxidant activities. It prevents a variety of chronic diseases by regulating fatty acid oxidation in the liver and gene expression in adipose tissue. It is also valuable in cancer and cardiovascular disease research.
  • Orally available bioactive component
  • Regulates fatty acid oxidation in the liver
  • Regulates gene expression in adipose tissue
  • Useful in cancer and cardiovascular disease research