Learn More
TARGETMOL CHEMICALS INC ISOFLUPREDONE 5MG
SDP
Catalog No. 502576802
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Isoflupredone (Isoflupredonum) is a synthetic corticosteroid that acts in vivo by binding to glucocorticoid and corticosteroid receptors in animals (such as horses) to inhibit inflammatory cells and inhibit the expression of inflammatory mediators. Isoflupredone has anti-inflammatory and immunosuppressive effects and can be used to treat a variety of musculoskeletal allergic and systemic inflammatory diseases. purity: 99%
Catalog No. 50-257-6802 Supplier TARGETMOL CHEMICALS INC Supplier No. T413035MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Isoflupredone (Isoflupredonum) is a synthetic corticosteroid that acts in vivo by binding to glucocorticoid and corticosteroid receptors in animals (such as horses) to inhibit inflammatory cells and inhibit the expression of inflammatory mediators. Isoflupredone has anti-inflammatory and immunosuppressive effects and can be used to treat a variety of musculoskeletal allergic and systemic inflammatory diseases. purity: 99%