Learn More
Braintree Scientific Isoflurane Vaporizer Anti-Spill Adapter
SDP
Catalog No. 501956097
Encompass
Isoflurane Funnel/Adaptor. Purple. This handy adapter screws onto gas agent bottle top and the other end fits snugly into the filling funnel on the gas vaporizer. It makes vaporizer refilling spill free. Color coded for agent type **Sold for research purposes only and not for human use**
This item is not returnable. View return policy
Catalog No. 50-195-6097 Supplier Braintree Scientific Supplier No. VPIANTISPILL
Isoflurane Funnel/Adaptor. Purple. This handy adapter screws onto gas agent bottle top and the other end fits snugly into the filling funnel on the gas vaporizer. It makes vaporizer refilling spill free. Color coded for agent type **Sold for research purposes only and not for human use**