Learn More
Stoelting™ Isoflurane Vaporizers
SDP
Catalog No. 10001092 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Pin index/key fill
Standard funnel fill
For Use With (Equipment):
Link 7 only
TECH 3 and Link 7
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model For Use With (Equipment)
10-001-092 Standard funnel fill TECH 3 and Link 7
10-000-928 Pin index/key fill Link 7 only
10-000-929 Standard funnel fill Link 7 only
10-001-091 Pin index/key fill TECH 3 and Link 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-001-092 Supplier Stoelting™ Supplier No. 539031
Add to Cart
Add to Cart
Stoelting™ Isoflurane Vaporizers are used on the Gas Anesthesia Systems to ensure that the output concentration is not affected by flow rate, temperature variation, duration of use, liquid level, or back pressure fluctuation.
  • Specifically calibrated for use with LINK 7
  • Extended capacity
  • 10 year warranty contingent upon a 2 year service cycle
  • Larger capacity for liquid anesthetic means filling the vaporizer less often which, in turn, reduces exposure to anesthetic gases.
  • Available with either funnel fill or pin index/key fill systems.
Specifications
For Use With (Application) Neuroscience
For Use With (Equipment) TECH 3 and Link 7
Size 250 mL
Model Standard funnel fill