Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isoformononetin | 486-63-5 | 10 MG
SDP

Catalog No. 5000418577
Encompass_Preferred

Isoformononetin is an analog of Daidzein with immunoprotective effects, inhibiting the differentiation of Th17 and B-cells lymphopoiesis to promote osteogenesis in estrogen-deficient bone loss conditions.

  • Molecular formula: C₁₆H₁₂O₄
  • Molecular weight: 268.26
  • Storage: 4°C, protected from light; in solvent: -80°C for 6 months, -20°C for 1 month (protected from light)
  • Solubility (in vitro): DMSO at 250 mg/mL (931.93 mM), requires ultrasonic
  • Usage caution: Product has not been fully validated for medical applications; for research use only

Catalog No. 50-004-18577 Supplier Medchemexpress LLC Supplier No. HYN750110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoformononetin is an analog of Daidzein with immunoprotective effects, inhibiting the differentiation of Th17 and B-cells lymphopoiesis to promote osteogenesis in estrogen-deficient bone loss conditions.

  • Molecular formula: C₁₆H₁₂O₄
  • Molecular weight: 268.26
  • Storage: 4°C, protected from light; in solvent: -80°C for 6 months, -20°C for 1 month (protected from light)
  • Solubility (in vitro): DMSO at 250 mg/mL (931.93 mM), requires ultrasonic
  • Usage caution: Product has not been fully validated for medical applications; for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.