Learn More
Medchemexpress LLC Isoformononetin | 486-63-5 | 5 MG
SDP
Catalog No. 5000419738
Encompass_Preferred
Isoformononetin is an analog of Daidzein with immunoprotective effects.
  • Inhibits the differentiation of Th17 cells
  • Inhibits B-cell lymphopoiesis
  • Promotes osteogenesis in estrogen-deficient bone loss conditions
Catalog No. 50-004-19738 Supplier Medchemexpress LLC Supplier No. HYN75015MG
Isoformononetin is an analog of Daidzein with immunoprotective effects.
  • Inhibits the differentiation of Th17 cells
  • Inhibits B-cell lymphopoiesis
  • Promotes osteogenesis in estrogen-deficient bone loss conditions