Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isoformononetin-d3 | 1 MG
SDP

Catalog No. 5000419647
Encompass_Preferred

Isoformononetin-d3 is a deuterium-labeled derivative of Isoformononetin, an isoflavone that functions as an analog of Daidzein. Isoformononetin has been noted for its immunoprotective effects. This compound has a molecular formula of C16H9D3O4 and a molecular weight of 271.28.

  • Deuterium-labeled compound
  • Analog of Daidzein
  • Exhibits immunoprotective effects
  • Molecular formula: C16H9D3O4
  • Molecular weight: 271.28

Catalog No. 50-004-19647 Supplier Medchemexpress LLC Supplier No. HYN7501S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoformononetin-d3 is a deuterium-labeled derivative of Isoformononetin, an isoflavone that functions as an analog of Daidzein. Isoformononetin has been noted for its immunoprotective effects. This compound has a molecular formula of C16H9D3O4 and a molecular weight of 271.28.

  • Deuterium-labeled compound
  • Analog of Daidzein
  • Exhibits immunoprotective effects
  • Molecular formula: C16H9D3O4
  • Molecular weight: 271.28

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.