Learn More
Medchemexpress LLC Isoforskolin | 64657-21-2 | 1 MG
SDP
Catalog No. 5000417783
Encompass_Preferred
Isoforskolin is the principal active component of C. forskohlii, native to China. It functions as an anti-inflammatory agent by reducing the secretion of lipopolysaccharide (LPS)-induced cytokines, specifically TNF-α, IL-1β, IL-6, and IL-8, in human mononuclear leukocytes. It is used for the treatment of Lyme arthritis.
  • Principal active component of C. forskohlii.
  • Reduces LPS-induced cytokine secretion (TNF-α, IL-1β, IL-6, and IL-8).
  • Acts as an anti-inflammatory agent.
  • Potentially treats Lyme arthritis.
Catalog No. 50-004-17783 Supplier Medchemexpress LLC Supplier No. HYN69271MG
Isoforskolin is the principal active component of C. forskohlii, native to China. It functions as an anti-inflammatory agent by reducing the secretion of lipopolysaccharide (LPS)-induced cytokines, specifically TNF-α, IL-1β, IL-6, and IL-8, in human mononuclear leukocytes. It is used for the treatment of Lyme arthritis.
  • Principal active component of C. forskohlii.
  • Reduces LPS-induced cytokine secretion (TNF-α, IL-1β, IL-6, and IL-8).
  • Acts as an anti-inflammatory agent.
  • Potentially treats Lyme arthritis.