Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isoforskolin | 64657-21-2 | 5 MG
SDP

Catalog No. 5000419936
Encompass_Preferred

Isoforskolin, also known as Coleonol B, is the principal active component derived from C. forskohlii, a plant native to China. This compound is recognized for its potent anti-inflammatory properties. It works by reducing the secretion of lipopolysaccharide (LPS)-induced cytokines, specifically targeting TNF-α, IL-1β, IL-6, and IL-8, in human mononuclear leukocytes. Research indicates its potential as an anti-inflammatory agent for treating conditions such as Lyme arthritis, where it has been shown to downregulate the transcription and expression of TNF-α and IL-6 induced by recombinant Borrelia burgdorferi basic membrane protein A (rBmpA).

  • Acts as an anti-inflammatory agent
  • Reduces LPS-induced cytokine secretion
  • Potential for treating Lyme arthritis
  • Purity of 98.0%
  • Molecular weight of 410.50
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • For research use only

Catalog No. 50-004-19936 Supplier Medchemexpress LLC Supplier No. HYN69275MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoforskolin, also known as Coleonol B, is the principal active component derived from C. forskohlii, a plant native to China. This compound is recognized for its potent anti-inflammatory properties. It works by reducing the secretion of lipopolysaccharide (LPS)-induced cytokines, specifically targeting TNF-α, IL-1β, IL-6, and IL-8, in human mononuclear leukocytes. Research indicates its potential as an anti-inflammatory agent for treating conditions such as Lyme arthritis, where it has been shown to downregulate the transcription and expression of TNF-α and IL-6 induced by recombinant Borrelia burgdorferi basic membrane protein A (rBmpA).

  • Acts as an anti-inflammatory agent
  • Reduces LPS-induced cytokine secretion
  • Potential for treating Lyme arthritis
  • Purity of 98.0%
  • Molecular weight of 410.50
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.