Learn More
Medchemexpress LLC Isoforsythiaside | 1357910-26-9 | 5 MG
SDP
Catalog No. 5000419039
Encompass_Preferred
Isoforsythiaside is an antioxidant and antibacterial phenylethanoid glycoside, exhibiting antibacterial activity against Escherichia coli, Pseudomonas aeruginosa, and Staphylococcus aureus. It is a glycoside isomer of Forsythiaside and is intended for research use only.
  • Antioxidant and antibacterial properties
  • Effective against various bacterial strains
  • Glycoside isomer
  • For research use only
Catalog No. 50-004-19039 Supplier Medchemexpress LLC Supplier No. HYN25945MG
Isoforsythiaside is an antioxidant and antibacterial phenylethanoid glycoside, exhibiting antibacterial activity against Escherichia coli, Pseudomonas aeruginosa, and Staphylococcus aureus. It is a glycoside isomer of Forsythiaside and is intended for research use only.
  • Antioxidant and antibacterial properties
  • Effective against various bacterial strains
  • Glycoside isomer
  • For research use only