Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isoforsythiaside | 1357910-26-9 | MFCD32645094 | 99.9% | 624.59 g/mol | C29H36O15 | 10 MG
SDP

Catalog No. 5000289508
Encompass_Preferred

Isoforsythiaside is an antioxidant and antibacterial phenylethanoid glycoside provided as a solid research compound. It exhibits activity against common bacterial strains and is supplied in small milligram packs for laboratory studies.

  • Antioxidant and antibacterial phenylethanoid glycoside with reported MICs: E. coli 40.83 μg/mL; P. aeruginosa 40.83 μg/mL; S. aureus 81.66 μg/mL.
  • High purity (99.94%).
  • Chemical formula C29H36O15 and molecular weight 624.59 g/mol.
  • Solid appearance, white to light yellow.
  • Recommended storage: 4°C sealed; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Supplied in small milligram packs suitable for research use.

Catalog No. 50-002-89508 Supplier Medchemexpress LLC Supplier No. HYN259410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoforsythiaside is an antioxidant and antibacterial phenylethanoid glycoside provided as a solid research compound. It exhibits activity against common bacterial strains and is supplied in small milligram packs for laboratory studies.

  • Antioxidant and antibacterial phenylethanoid glycoside with reported MICs: E. coli 40.83 μg/mL; P. aeruginosa 40.83 μg/mL; S. aureus 81.66 μg/mL.
  • High purity (99.94%).
  • Chemical formula C29H36O15 and molecular weight 624.59 g/mol.
  • Solid appearance, white to light yellow.
  • Recommended storage: 4°C sealed; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Supplied in small milligram packs suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.