Learn More
TARGETMOL CHEMICALS INC ISOFRAXIDIN 25MG
SDP
Catalog No. 502579722
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. 1. Isofraxidin (Phytodolor) protects leukemia cells from radiation-induced apoptosis via ROS/mitochondria pathway in a p53-independent manner. 2. Isofraxidin has a protective effect against LPS-induced ALI and the protective effect of Isofraxidin seems to result from the inhibition of COX-2 protein expression in the lung which regulates the production of PGE2. 3. Isofraxidin possesses significant analgesic and anti-inflammatory activities that may be mediated through the regulation of pro-inflammatory cytokines TNF-(alpha) and the phosphorylation of p38 and ERK1/2. purity: 99%
Catalog No. 50-257-9722 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S004525MG
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. 1. Isofraxidin (Phytodolor) protects leukemia cells from radiation-induced apoptosis via ROS/mitochondria pathway in a p53-independent manner. 2. Isofraxidin has a protective effect against LPS-induced ALI and the protective effect of Isofraxidin seems to result from the inhibition of COX-2 protein expression in the lung which regulates the production of PGE2. 3. Isofraxidin possesses significant analgesic and anti-inflammatory activities that may be mediated through the regulation of pro-inflammatory cytokines TNF-(alpha) and the phosphorylation of p38 and ERK1/2. purity: 99%