Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isofraxidin | 486-21-5 | 222.19 | 5 MG
SDP

Catalog No. 5000416979
Encompass_Preferred

Isofraxidin, a coumarin component from *Acanthopanax senticosus*, inhibits MMP-7 expression and cell invasion of human hepatoma cells. It inhibits the phosphorylation of ERK1/2 in hepatoma cells and attenuates the expression of iNOS and COX-2. Additionally, Isofraxidin inhibits TLR4/myeloid differentiation protein-2 (MD-2) complex formation.

  • Inhibits MMP-7 expression and cell invasion
  • Inhibits phosphorylation of ERK1/2
  • Attenuates expression of iNOS and COX-2
  • Inhibits TLR4/MD-2 complex formation
  • Purity: 99.94%
  • Appearance: Solid, off-white to yellow color
  • Soluble in DMSO: 250 mg/mL

Catalog No. 50-004-16979 Supplier Medchemexpress LLC Supplier No. HYN07745MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isofraxidin, a coumarin component from *Acanthopanax senticosus*, inhibits MMP-7 expression and cell invasion of human hepatoma cells. It inhibits the phosphorylation of ERK1/2 in hepatoma cells and attenuates the expression of iNOS and COX-2. Additionally, Isofraxidin inhibits TLR4/myeloid differentiation protein-2 (MD-2) complex formation.

  • Inhibits MMP-7 expression and cell invasion
  • Inhibits phosphorylation of ERK1/2
  • Attenuates expression of iNOS and COX-2
  • Inhibits TLR4/MD-2 complex formation
  • Purity: 99.94%
  • Appearance: Solid, off-white to yellow color
  • Soluble in DMSO: 250 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.