Learn More
Alkali Scientific IsoFreeze Flipper Racks with Lid, 1 Rack/Unit, Subzero, Blue, 20°C
SDP
Catalog No. 5000661971
Encompass_Preferred
IsoFreeze™ temperature maintenance racks provide safe chilled and stable storage for PCR reagents, cells, enzymes or other temperature sensitive reagents when removed from the freezer or refrigerator for extended periods of time. 20 wells in 4 x 5 format. Consistent thermal protection. Standard IsoFreeze Flipper Rack, Maintains 0°C on bench top for minimum of 5 hours Subzero IsoFreeze Flipper Rack, Maintains 15°C to 20°C on bench top for minimum of 3 hours One side holds 20 x 1.5/2.0 mL tubes, and the other side holds 20 x 0.5 mL tubes. Tough polycarbonate material. Racks are filled with an inert, non toxic gel. Lid included.
Catalog No. 50-006-61971 Supplier Alkali Scientific Supplier No. ISFB020
IsoFreeze™ temperature maintenance racks provide safe chilled and stable storage for PCR reagents, cells, enzymes or other temperature sensitive reagents when removed from the freezer or refrigerator for extended periods of time. 20 wells in 4 x 5 format. Consistent thermal protection. Standard IsoFreeze Flipper Rack, Maintains 0°C on bench top for minimum of 5 hours Subzero IsoFreeze Flipper Rack, Maintains 15°C to 20°C on bench top for minimum of 3 hours One side holds 20 x 1.5/2.0 mL tubes, and the other side holds 20 x 0.5 mL tubes. Tough polycarbonate material. Racks are filled with an inert, non toxic gel. Lid included.