Learn More
Braintree Scientific IsoFreeze PCR Rack
SDP
Catalog No. 501954976
Encompass
IsoFreeze PCR RackConsistently cooled samples the new IsoFreeze PCR Racks When preparing PCR samples in plates, strips and single tubes, PCR solutions must be kept cool as warming may result in unspecific PCR products. Our new IsoFreeze PCR Racks prevent sample damage caused by premature warming. Using IsoFreeze PCR Racks provides considerable advantages: Once pre-chilled, the rack can maintain the PCR sample temperature at approximately 4C for up to 3 hours in ambient conditions. If sample temperatures exceed 7C, the rack visibly changes color from purple to pink to alert users. The racks are sold in a pack of two so that a warm rack can be conveniently exchanged for a chilled rack during prolonged use. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4976 Supplier Braintree Scientific Supplier No. ISOFREEZE
IsoFreeze PCR RackConsistently cooled samples the new IsoFreeze PCR Racks When preparing PCR samples in plates, strips and single tubes, PCR solutions must be kept cool as warming may result in unspecific PCR products. Our new IsoFreeze PCR Racks prevent sample damage caused by premature warming. Using IsoFreeze PCR Racks provides considerable advantages: Once pre-chilled, the rack can maintain the PCR sample temperature at approximately 4C for up to 3 hours in ambient conditions. If sample temperatures exceed 7C, the rack visibly changes color from purple to pink to alert users. The racks are sold in a pack of two so that a warm rack can be conveniently exchanged for a chilled rack during prolonged use. **Sold for research purposes only and not for human use**