Learn More
Andwin Scientific ISOFREEZE PCR SBS RACK 2PK
SDP
Catalog No. 50999415
Encompass_Preferred
The GeneMate IsoFreeze PCR SBS Racks can maintain a sub 4C (refrigerator) environment for up to 4 hours. This product is ideal for use with PCR reagents, cells, enzymes, or other temperature-sensitive reagents. The racks are able fit standard format 0.2 mL tube, strip tubes, and any format of PCR plate (up to 96 wells) for PCR, QPCR, or other applications. This is sold in a pack of 2 purple racks that changes its color to pink in warm temperature.
Catalog No. 50-999-415 Supplier Andwin Scientific Supplier No. R2023P
The GeneMate IsoFreeze PCR SBS Racks can maintain a sub 4C (refrigerator) environment for up to 4 hours. This product is ideal for use with PCR reagents, cells, enzymes, or other temperature-sensitive reagents. The racks are able fit standard format 0.2 mL tube, strip tubes, and any format of PCR plate (up to 96 wells) for PCR, QPCR, or other applications. This is sold in a pack of 2 purple racks that changes its color to pink in warm temperature.