Learn More
Research Products International Corp IsoFreeze Micro-Tube Chillers, Green/Yellow Indicator, 1.5 - 2.0 ml Tubes, 24 Tube Capacity
SDP
Catalog No. 50550286
Encompass
IsoFreeze chilling racks provide a stable low temperature environment for 24 x 1.5-2.0ml micro-tubes. Gel filled racks maintain a sample temperature of less than 4°C for up to four hours without refrigeration. Unique color change plastic indicates temperature at a glance.
Catalog No. 50-550-286 Supplier Research Products International Corp Supplier No. 248012
Add to Cart
Add to Cart
IsoFreeze chilling racks provide a stable low temperature environment for 24 x 1.5-2.0ml micro-tubes. Gel filled racks maintain a sample temperature of less than 4°C for up to four hours without refrigeration. Unique color change plastic indicates temperature at a glance.