Learn More
AdipoGen Isofusidienol A
SDP
Catalog No. 502562021
Encompass_Preferred
Chemical. CAS 1032392-18-9. Formula C16H12O6. MW 300.3. Isolated from Chalara sp. Antibiotic. Chromone derivative. Antifungal and antibacterial against Gram-positive and Gram-negative bacteria.
Catalog No. 50-256-2021 Supplier AdipoGen Supplier No. BVT0280M001
Chemical. CAS 1032392-18-9. Formula C16H12O6. MW 300.3. Isolated from Chalara sp. Antibiotic. Chromone derivative. Antifungal and antibacterial against Gram-positive and Gram-negative bacteria.