Learn More
Cayman Chemical Isogarcinol, Purity: ≥95%, CAS: 71117-97-0, 10 mg
SDP
Catalog No. 502983611
Encompass_Preferred
A natural polyisoprenylated benzophenone that has immunosuppressant actions in mouse models of inflammatory or autoimmune disease; inhibits the proliferation of HL-60 and PC-3 cancer cells (IC50s = 4 and 8 µg/ml, respectively) through cell cycle arrest and apoptosis
Catalog No. 50-298-3611 Supplier Cayman Chemical Supplier No. 2116410 MG
A natural polyisoprenylated benzophenone that has immunosuppressant actions in mouse models of inflammatory or autoimmune disease; inhibits the proliferation of HL-60 and PC-3 cancer cells (IC50s = 4 and 8 µg/ml, respectively) through cell cycle arrest and apoptosis