Learn More
IsoGel™ Agarose
Catalog No. BMA50202 Shop All Lonza, Inc Products
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
BMA50202 25 g
1 options
This item is not returnable. View return policy
Catalog No. BMA50202 Supplier Lonza Inc Supplier No. 50202
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • For isoelectric focusing (IEF)
  • No measurable EEO
  • Sufficiently rigid to be cast in vertical tubes
  • Gels cast on GelBond™ Film dry down quickly and easily to transparent permanent records
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cambrex Iso-Gel™ Agarose
Quantity 25 g