Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical isoGNA O6-DPC-G(iBu) amidite | 1 g | CAS 1638739-09-9 | InChIKey CNQFTLNOLDNADK-IKGLYBOLSA-N

Catalog No. 502365253
Encompass

isoGNA O6-DPC-G(iBu) amidite is a Amidite reagent (Subcategory: isoGNA Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 1638739-09-9
- MDL: No data
- InChIKey: CNQFTLNOLDNADK-IKGLYBOLSA-N
- Molecular Weight: 993.114761998
- Molecular Formula: C55H61N8O8P
- Purity: ≥98%
- Container Type: 30 mL Glass (20-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 47.3 g
- Commodity Code: 29335995
- Country Of Origin: China
- IUPAC: 9-((2S)-1-(bis(4-methoxyphenyl)(phenyl)methoxy)-3-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)propan-2-yl)-2-isobutyramido-9H-purin-6-yl diphenylcarbamate
- SMILES: CC(N(P(OC[C@H](COC(C1=CC=C(C=C1)OC)(C2=CC=C(C=C2)OC)C3=CC=CC=C3)N4C5=C(N=C4)C(OC(N(C6=CC=CC=C6)C7=CC=CC=C7)=O)=NC(NC(C(C)C)=O)=N5)OCCC#N)C(C)C)C

Catalog No. 50-236-5253 Supplier STA Pharmaceutical Supplier No. STAAM10631KA301GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

isoGNA O6-DPC-G(iBu) amidite is a Amidite reagent (Subcategory: isoGNA Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 1638739-09-9
- MDL: No data
- InChIKey: CNQFTLNOLDNADK-IKGLYBOLSA-N
- Molecular Weight: 993.114761998
- Molecular Formula: C55H61N8O8P
- Purity: ≥98%
- Container Type: 30 mL Glass (20-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 47.3 g
- Commodity Code: 29335995
- Country Of Origin: China
- IUPAC: 9-((2S)-1-(bis(4-methoxyphenyl)(phenyl)methoxy)-3-(((2-cyanoethoxy)(diisopropylamino)phosphaneyl)oxy)propan-2-yl)-2-isobutyramido-9H-purin-6-yl diphenylcarbamate
- SMILES: CC(N(P(OC[C@H](COC(C1=CC=C(C=C1)OC)(C2=CC=C(C=C2)OC)C3=CC=CC=C3)N4C5=C(N=C4)C(OC(N(C6=CC=CC=C6)C7=CC=CC=C7)=O)=NC(NC(C(C)C)=O)=N5)OCCC#N)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.