Learn More
Sigma Aldrich Fine Chemicals Biosciences ISOGROR 13C Powder Growth1G
Catalog No. 502905320
Encompass
ISOGRO(R) media is required for overcoming the growth limitations of minimal media. ISOGRO products are lysates of algae grown with stable isotopes (13C 15N and/or D). ISOGRO(R)-13C powder -growth medium gives uniform labeling for protein expression NMR (nuclear magnetic resonance) studies.A typical algal lysate (ISOGRO medium) contains 30% salts 3% water 2% glucose and 65% amino acids/peptides.
Catalog No. 50-290-5320 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6068631G
ISOGRO(R) media is required for overcoming the growth limitations of minimal media. ISOGRO products are lysates of algae grown with stable isotopes (13C 15N and/or D). ISOGRO(R)-13C powder -growth medium gives uniform labeling for protein expression NMR (nuclear magnetic resonance) studies.A typical algal lysate (ISOGRO medium) contains 30% salts 3% water 2% glucose and 65% amino acids/peptides.