Learn More
Medchemexpress LLC HY-100810 25mg Medchemexpress, Isoguvacine (hydrochloride) CAS:68547-97-7 Purity:>98%
SDP
Catalog No. 501931958
Encompass_Preferred
Medchemexpress, HY-100810 25mg Isoguvacine (hydrochloride) CAS:68547-97-7 Isoguvacine hydrochloride is a GABA receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1958 Supplier Medchemexpress LLC Supplier No. HY10081025MG
Medchemexpress, HY-100810 25mg Isoguvacine (hydrochloride) CAS:68547-97-7 Isoguvacine hydrochloride is a GABA receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.