Learn More
Medchemexpress LLC Isoguvacine (hydrochloride) | 68547-97-7 | 99.8% | 163.60 | C6H10ClNO2 | 1 ML
SDP
Catalog No. 5000213405
Encompass_Preferred
Isoguvacine hydrochloride is a GABAA receptor agonist used in neuroscience and pharmacology research to activate GABA receptors. It is supplied as research-grade material, available as a ready-to-use 10 mM stock solution in DMSO or as solid material for custom stock preparation.
  • Ready-to-use 10 mM stock solution in DMSO for rapid experimental setup.
  • Also available as solid for preparation of custom concentrations.
  • High reported purity (~99.8%) suitable for research applications.
  • Manufacturer-provided solubility and stock preparation guidance for water and DMSO.
  • Storage recommendations listed for both solvent and solid states to maintain stability.
Catalog No. 50-002-13405 Supplier Medchemexpress LLC Supplier No. HY10081010MM 1ML
Isoguvacine hydrochloride is a GABAA receptor agonist used in neuroscience and pharmacology research to activate GABA receptors. It is supplied as research-grade material, available as a ready-to-use 10 mM stock solution in DMSO or as solid material for custom stock preparation.
  • Ready-to-use 10 mM stock solution in DMSO for rapid experimental setup.
  • Also available as solid for preparation of custom concentrations.
  • High reported purity (~99.8%) suitable for research applications.
  • Manufacturer-provided solubility and stock preparation guidance for water and DMSO.
  • Storage recommendations listed for both solvent and solid states to maintain stability.