Learn More
Cayman Chemical Isoguvacine (hydrochloride), Purity: ≥95%, CAS: 68547-97-7, 100 mg
SDP
Catalog No. 502950312
Encompass_Preferred
A GABAA; binds to rat synaptic cortical membranes (Ki = 55 nM); activates α1β2γ2S, α2β2γ2S, α3β2γ2S, α5β2γ2S, and ρ1 subunit-contanining GABAA receptors in a FLIPR assay (EC50s = 13, 4.5, 5.6, 0.78, and 3.1 µM, respectively); inhibits CA1 population spikes in rat hippocampal slices (EC50 = 13 µM); inhibits potassium/low magnesium-induced seizures in rat pups; chronic administration improves tactile over-reactivity, anxiety-like behaviors, and social impairments in mouse models of ASD at 2 mg/kg
Catalog No. 50-295-0312 Supplier Cayman Chemical Supplier No. 33143100 MG
A GABAA; binds to rat synaptic cortical membranes (Ki = 55 nM); activates α1β2γ2S, α2β2γ2S, α3β2γ2S, α5β2γ2S, and ρ1 subunit-contanining GABAA receptors in a FLIPR assay (EC50s = 13, 4.5, 5.6, 0.78, and 3.1 µM, respectively); inhibits CA1 population spikes in rat hippocampal slices (EC50 = 13 µM); inhibits potassium/low magnesium-induced seizures in rat pups; chronic administration improves tactile over-reactivity, anxiety-like behaviors, and social impairments in mouse models of ASD at 2 mg/kg