Learn More
Boca Scientific Inc Buccalyse Rapid Buccal Cell DNA Isolation Kit, 50 Extractions, with 50 SK-1S Buccal Swabs
Catalog No. NC1870693
Encompass
The BEK-50/SK1 kit includes 50 SK-1S Buccal Swabs and the Buccalyse DNA extraction kit for 50 samples. SK-1S Buccal Swabs are designed for high-yield DNA retrieval and convenient transport. Each SK-1S buccal swab is packaged with a 5ml plastic collection tube and cap. The Buccalyse DNA Extraction Kit, specifically designed for use with buccal swabs, includes one solution for a fast and simple one tube method for extracting DNA from buccal swabs. In only 20 minutes, this kit provides PCR-ready DNA that does not require centrifugation.
This item is not returnable. View return policy
Catalog No. NC1870693 Supplier Boca Scientific Inc Supplier No. BEK50/SK1
The BEK-50/SK1 kit includes 50 SK-1S Buccal Swabs and the Buccalyse DNA extraction kit for 50 samples. SK-1S Buccal Swabs are designed for high-yield DNA retrieval and convenient transport. Each SK-1S buccal swab is packaged with a 5ml plastic collection tube and cap. The Buccalyse DNA Extraction Kit, specifically designed for use with buccal swabs, includes one solution for a fast and simple one tube method for extracting DNA from buccal swabs. In only 20 minutes, this kit provides PCR-ready DNA that does not require centrifugation.