Learn More
Boca Scientific Inc Isohelix DNA/RNA Buccal Swabs, wrapped individually, without collection tubes, pack of 250
Catalog No. NC1894506
Encompass
Isohelix buccal swabs have been specifically designed to offer increased yields of high quality buccal cell and genomic DNA and all species of RNA. Buccal cell sampling offers a viable alternative to blood collection, and together with the Isohelix range of Isolation and Stabilization kits, enables high yields of pure, intact DNA and RNA to be collected easily and quickly. Our swabs are suitable for both human and veterinary use. The swab design offers significant advantages in efficiency of cell collection with the unique swab matrix, which in combination with a quick release surface maximizes DNA and RNA yields. Swab batches are ethylene oxide treated and routinely tested for human contamination. Each SK-3S buccal swab is wrapped individually. The swabs include a breakpoint to quickly snap the swab head off the shaft if you wish to store the swab in a tube. Collection tubes are not included.
This item is not returnable. View return policy
Catalog No. NC1894506 Supplier Boca Scientific Inc Supplier No. SK3S
Isohelix buccal swabs have been specifically designed to offer increased yields of high quality buccal cell and genomic DNA and all species of RNA. Buccal cell sampling offers a viable alternative to blood collection, and together with the Isohelix range of Isolation and Stabilization kits, enables high yields of pure, intact DNA and RNA to be collected easily and quickly. Our swabs are suitable for both human and veterinary use. The swab design offers significant advantages in efficiency of cell collection with the unique swab matrix, which in combination with a quick release surface maximizes DNA and RNA yields. Swab batches are ethylene oxide treated and routinely tested for human contamination. Each SK-3S buccal swab is wrapped individually. The swabs include a breakpoint to quickly snap the swab head off the shaft if you wish to store the swab in a tube. Collection tubes are not included.