Learn More
Selleck Chemical LLC Isohomovanillic acid
SDP
Catalog No. 5000003962
Encompass_Preferred
Isohomovanillic acid is a deaminated metabolite of catecholamines formed by the enzyme catechol-O-methyltransferase which catalyzes the transfer of a methyl group from S-adenosylmethionine to catecholamines including the neurotransmitters dopamine epinephrine and norepinephrine
Catalog No. 50-000-03962 Supplier Selleck Chemical LLC Supplier No. S647925MG
Add to Cart
Add to Cart
Isohomovanillic acid is a deaminated metabolite of catecholamines formed by the enzyme catechol-O-methyltransferase which catalyzes the transfer of a methyl group from S-adenosylmethionine to catecholamines including the neurotransmitters dopamine epinephrine and norepinephrine