Learn More
Medchemexpress LLC Isohomovanillic acid | 1131-94-8 | 182.18 | 1 ML
SDP
Catalog No. 5000446926
Encompass_Preferred
Isohomovanillic acid (3-Hydroxy-4-methoxyphenylacetic acid) is a crucial product of catecholamine metabolism, naturally found in urine. It serves as a valuable compound in Parkinson's disease research. This high-purity chemical has a molecular weight of 182.18 and a molecular formula of C9H10O4, making it suitable for various laboratory applications.
  • High purity (99.12%) for reliable research outcomes
  • Effective in Parkinson's disease research applications
  • Soluble in DMSO and water for flexible experimental setups
  • Stable when stored properly in powder or solution form
Catalog No. 50-004-46926 Supplier Medchemexpress LLC Supplier No. HYW00211010MM/1ML
Add to Cart
Add to Cart
Isohomovanillic acid (3-Hydroxy-4-methoxyphenylacetic acid) is a crucial product of catecholamine metabolism, naturally found in urine. It serves as a valuable compound in Parkinson's disease research. This high-purity chemical has a molecular weight of 182.18 and a molecular formula of C9H10O4, making it suitable for various laboratory applications.
  • High purity (99.12%) for reliable research outcomes
  • Effective in Parkinson's disease research applications
  • Soluble in DMSO and water for flexible experimental setups
  • Stable when stored properly in powder or solution form