Learn More
Cayman Chemical IsoImperatrIn 10mg
SDP
Catalog No. 502983791
Encompass_Preferred
A natural furanocoumarin that inhibits CYP1A activity, in particular blocking hepatic EROD activity; also induces the expression of GSTα by activating Nrf2
Catalog No. 50-298-3791 Supplier Cayman Chemical Supplier No. 1985110 MG
A natural furanocoumarin that inhibits CYP1A activity, in particular blocking hepatic EROD activity; also induces the expression of GSTα by activating Nrf2